Coomassie Stain, Bio-Safe™, Bio-Rad
供应商: Bio-Rad
Bio-Safe™
BRAD161-0786EA 0 CNY
BRAD161-0786 BRAD161-0787
Coomassie Stain, Bio-Safe™, Bio-Rad
染料 电泳污渍
Premixed staining solution, for polyacrylamide protein gels. Choose the flexibility of Bio-Safe Coomassie Premixed Staining Solution. Proteins can be visualized in polyacrylamide gels with or without the use of acetic acid/methanol fixation.
  • No requirement for fixation* with acetic acid and methanol
  • Destain with water
  • No handling and disposal of solvents
  • Gels can be stored in water
* Fixation is recommended for peptide and very low molecular weight protein separations prior to using Bio-Safe Coomassie Stain.