VWR®Standard Series, Manual defrost laboratory freezers
供应商: VWR
VWR®
VWRU10819-410EA 0 CNY
VWRU10819-410
VWR®Standard Series, Manual defrost laboratory freezers
冷柜 实验室通用冰箱
These standard series manual defrost laboratory freezers are designed specifically with general-purpose applications in mind.
  • Adjustable operating temperature range from –15 to –25 °C
  • Direct-cool fixed evaporator shelves
  • Provides freezing without precise temperature control
  • Supplied with two-year parts and labour warranty, and five years warranty on compressor parts
These units are ideal when stable temperatures averaging –20 °C are required but precise temperature control, maintenance, and alarms are not necessary. These units feature a manual defrost design to eliminate damaging temperature spikes from defrost cycles. This ensures valuable sample security. Defrosting of the unit must be performed periodically to prevent severe ice build-up.
Certifications: UL/C-UL listed.