VWR® Performance Series, Solid door stainless steel auto defrost laboratory freezer with natural refrigerant
供应商: VWR
VWR®
VWRU10819-492EA 0 CNY
VWRU10819-492
VWR® Performance Series, Solid door stainless steel auto defrost laboratory freezer with natural refrigerant
冷柜 实验室通用冰箱
Freezers feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.
  • Data acquisition
  • 24/7 data logging
  • 24-hour digital temp chart
  • Min/max temp history
  • Swivel casters (adds 127 mm to height; included in dimensions)
A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings.
Features keyed door locks, casters, sample-simulated probe, probe access port, multi-point temperature mapping via NIST traceable data acquisition, 15 probes in air and 3 probes in sample solution for 360 minutes with temperature readings every 60 seconds.
Certifications: UL/C-UL listed.
订购须知 Units are supplied with a three-year parts and labour warranty, plus an additional two-year compressor parts warranty.