VWR® Basic Protection face masks feature a two-ply construction with an ultra-soft inner layer designer to provide a high level of comfort.
- Latex-free
- Protects against bacteria and particulates
- High breathability and fluid-resistance
- Soft and hypoallergenic
Ultrasonically welded spandex ear loops and malleable 12 cm aluminium noseband create a secure facial seal and improve wearer comfort.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Validated through independent laboratory testing.
Packaging: 50 masks per pack, 10 packs per case.