Compact anemometer measures air velocity and temperature with simple, one-hand operation. The instrument displays air velocity in metres per second, feet per second, miles per hour, kilometres per hour, and knots.
- Displays air velocity, max. air velocity measured, temperature, wind chill temperature
- Tough, water and shock resistant ABS plastic housing
- Temperature in °C and °F
- Easy to operate – ideal for use in hoods or outdoors
- Extra-large 13 mm LCD digits are legible from 3 m
Readings are updated instantaneously. The memory function recalls highest and lowest readings. The hold key freezes the display for capturing a reading in a difficult location. The average key automatically calculates mean value for up to eight readings.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery information: Supplied with RS-232 serial output jack, 9 V alkaline battery, instructions, and carrying case.