VWR® Polyclean algaecide Water bath protective agent
供应商: Avantor
VWR®
VWRU71002-500EA 0 CNY
VWRU71002-500
VWR® Polyclean algaecide Water bath protective agent
清洗剂 水浴保护剂
Prevents growth of algae, keeps baths clean and odour-free.
Bottle of 237 ml solution treats approximately 757 litres of water.
Suitable for use with VWR® refrigerated circulating baths, VWR® heated circulating baths and VWR® circulating open bath systems.