Designed for in situ hybridisation, slide warming and drying, immuno-histochemistry, FISH, and all single-temperature microscope slide incubation procedures.
- Excellent temperature uniformity and stability
- Temperature stability: ±0,2 °C
- Temperature uniformity: ±0,35 °C at 37 °C, ±0,07 °C at 65 °C
- Easy to insert and remove slides
- Bath heats up quickly
Bath features a PID controller for easy temperature setting, and exhibits excellent temperature uniformity and stability. This device holds up to 30 standard microscope slides, divided into three rows of 10. The clearance between lid and slide pedestal is 10 mm. The bath heats up from ambient to 65 °C (149 °F) in 20 minutes.
订购须知 VWR® two-year limited parts and labor warranty.
Delivery information: Supplied with a tempered glass lid that seals via an integral gasket on the heating base.