VWR®, Carboy, Narrow Mouth, Round, Sterile
供应商: VWR
VWR®
VWRU75795-776EA 0 CNY
VWRU75795-776
VWR®, Carboy, Narrow Mouth, Round, Sterile
carboy水瓶
Use for storing and dispensing solutions and media. Ideal for sterile water storage. These disposable carboys are ideal for storing and transfer of sterile fluids and pharmaceutical/biotech reagents.
  • Sterile
  • Excellent chemical resistance
  • Clear graduation marks
  • Narrow-mouth
  • Integrated shoulder handle
They are well suited for use in temperatures ranging from –100 to 100 °C (–148 to 212 °F). Carboys are leak-proof under standard test conditions.
They are not autoclavable.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.