VWR®, Protection covers, chemical resistant Maximum
供应商: VWR
VWR® Maximum
VWRU414004-461EA 0 CNY
VWRU414004-461
VWR®, Protection covers, chemical resistant Maximum
保护罩
These chemical-resistant covers provide the highest level of protection available within the VWR® Protection line.
  • Superior chemical repellence as well as liquid and particle holdout
  • Double-coated fabric protects against hazardous liquids and toxic chemicals
  • Maximum durability and tear strength
Engineered to optimise barrier protection and prevent penetration by an extensive list of chemicals.
Delivery information: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.