The Standard Model 1000 shaker combines the size and economic benefits of a mini-shaker with the orbit and speed of a more traditional unit.
- Low-profile design
- Variable speed microprocessor control
- Can be used in cold rooms, incubators, and CO2 environments from 0 to 40 °C (32 to 104 °F).
The space-saving, low-profile design easily fits into most fume hoods and incubators.
Shakers are equipped with a ramping feature that slowly increases speed to the desired setpoint to avoid splashing or sample loss. Units feature a spill-resistant design that channels fluids away from internal components.
A cast aluminum base offers durability and added stability. The variable speed microprocessor control provides consistent uniform mixing action, and the powerful triple eccentric drive and brushless motor ensure reliable service in continuous duty. A basic speed knob with 1 to 10 dial settings controls all adjustments.
Certifications: RoHS certified. CSA certified.
订购须知 Supplied with a non skid rubber mat for securing Petri dishes and cell culture flasks. Also included is a 2,3 mm (92") detachable, 3-wire cord and plug for 120 V model.