VWR®, Meker Burner, High Temperature
供应商: VWR
VWR®
VWRU89038-548EA 0 CNY
VWRU89038-548
VWR®, Meker Burner, High Temperature
加热器 煤气灯
Meker high-temperature burner which is ideal for use where steady, high temperatures are required.
  • Backflashes eliminated via safety cap feature
  • Short, wide, uniform flame
  • Micrometer needle valve
Special micrometer needle valve regulates gas. It features injector-shaped 2,9 mm venturi tube.
Burner head dimension (Ø×H): 10×203 mm.