VWR®, Test Tube Baskets, Epoxy Coated Aluminium
供应商: VWR
VWR®
VWRU89259-810EA 0 CNY
VWRU89259-810
VWR®, Test Tube Baskets, Epoxy Coated Aluminium
篮筐 试管筐
Aluminium, epoxy coated test tube baskets are using for holding laboratory ware for washing, storage, or autoclaving.
  • Flow-through design
  • Durable under many laboratory environments
  • Withstands broad temperature ranges from –220 to 120 °C
Colour: White