VWR®, Spoon Spatulas, Porcelain
供应商: VWR
VWR®
VWRU89038-208EA 0 CNY
VWRU89038-208 VWRU89038-206
VWR®, Spoon Spatulas, Porcelain
样品勺
Chemical-porcelain spatulas feature a spatula on one end and a spoon on the other.
  • High chemical resistance
  • Glazed for added protection