VWR®, Scoops, PE, Cylindrical
供应商: VWR
VWR®
VWRU70680-280EA 800 CNY
VWRU70680-280 VWRU36494-110
VWR®, Scoops, PE, Cylindrical
样品铲
Large, heavy-duty, polyethylene plastic scoops are strong, durable and excellent for scooping dry powders, granules, or crystals.
  • Closed back, non-spill shape
  • Ergonomically angled handle
Handle length: 127 mm