The VWR® universal homogenising package is designed for use with all types of samples, even tough or stringy.
- Fast and efficient operation
- Flexibility of homogenising within a wide range of tube sizes, from micro-tubes to large tubes (1,5 - 50 ml tubes)
- Stand allows hands free use
The 7 mm and 10 mm saw-tooth generator probes provide fast and efficient operation. From soft to tough samples, both saw-tooth generator probes provide extra cutting action during homogenising, ensuring fast and complete sample breakdown.
Delivery information: Package includes VWR® 200 homogeniser unit, 7×95 mm saw-tooth generator probe, 10×115 mm saw-tooth generator probe, and stand assembly.