VWR®, Rubber Stopper Assortment, Black
供应商: VWR
VWR®
VWRU59582-585EA 0 CNY
VWRU59582-585
VWR®, Rubber Stopper Assortment, Black
塞子 橡胶塞
This two-pound assortment of black rubber stoppers is ideal for any small laboratory or school with limited need for any one size or style stopper.
  • Practical
  • 66 pieces
Assortment consists of solid, one-hole and two-hole stoppers, sizes 00 through 8.