VWR®, Undercounter Glassware Washers
供应商: VWR
VWR®
VWRU10002-854EA 0 CNY
VWRU10002-854
VWR®, Undercounter Glassware Washers
玻璃器皿洗涤机
Economical glassware washers feature stainless steel construction for long life and corrosion protection.
  • Stainless steel construction ensures long life and corrosion protection
  • Insulated external casing
  • Designed for optimum heat and noise absorption
  • Three separate filters protect pumps from debris
  • Multiple cycle selections and microprocessor control systems
  • Enhanced dual fan drying system
  • Temperature selection to 70 °C (158 °F) for optimal cleaning of different loads
  • Delayed start feature
With adjustable legs, these washers can fit into any standard cabinet. New improved units feature a soft touch controller, improved drying, a quieter run, and are compatible with existing racks and accessories. Engineered for reliable and energy efficient operation.
Certifications: ETL and ETL-C listed.
订购须知 A wide range of accessories are available to fit both upper and lower racks; spindle racks, specialty baskets, and other inserts. For more information, contact your local Avantor sales office.