盐酸胍

供应商: INVITROGEN
Warning

Synonyms: 亚氨脲盐酸胍亚氨基脲

INVT15502016EA 0 CNY
INVT15502016
盐酸胍
盐酸胍
分子式: NH₂C(=NH)NH₂·HCl
分子量: 95,53 g/mol
溶点: 185 °C
密度: 1,354 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00013026
CAS号: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://cn.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR