Automated cell counter, Vi-CELL XR
供应商: Beckman Coulter
BECL383556EA 0 CNY
BECL383556
Automated cell counter, Vi-CELL XR
细胞计数器 电子细胞计数器
Vi-CELL automates the manual trypan blue viability method and removes the subjective variables inherent in the manual assay.
  • Reports cell concentration, size distribution and circularity measurements
  • Real time cell images can be archived for future analyses