Handheld turbidity and chlorine meter
供应商: Hanna
HANNHI93414EA 0 CNY
HANNHI93414
Handheld turbidity and chlorine meter
浊度计
Designed for water quality measurements on low level turbidity and chlorine samples.
  • Less stray light and more accuracy at low ranges
  • USB and RS232 PC connectivity for download of data
  • Backlit LCD with user friendly display and guidance codes
  • GLP functions for traceability of calibration conditions including time, date and last calibration points
  • Fast Tracker™ tag identification system for data management as well as chlorine CAL Check™ calibration validation
Certifications: EPA compliant Tungsten light source.
Delivery information: Supplied with five cuvettes and caps, calibration cuvettes, silicone oil, cleaning tissues, tag holders with tags, scissors, four batteries, AC adapter, instruction manual, quality certificate and carrying case.