VWR® Premium, Disposable microtome blade
供应商: VWR
VWR®
VWRU95057-832EA 0 CNY
VWRU95057-834 VWRU95057-832
VWR® Premium, Disposable microtome blade
切片机 Microtome Blades and Handles
Designed to fit standard blade holders, premium disposable blades are ideal for all routine and speciality histology applications.
  • Heavy gauge high-quality carbon steel construction
  • Ultra-thin sectioning
  • Sections jump off edge forming beautiful ribbons
Low-profile triple-facet blades feature an extra-sharp cutting edge for thinner sections that will cut down to 2 µm, and a fluoropolymer resin coating to ensure a consistent cutting edge and increase blade life.
Thick high-profile blades eliminate chattering when sectioning hard specimens. This blade will not bend during cutting, eliminating the thick/thin sectioning problem often found when cutting hard tissue. These blades are ideal for sectioning bone, eyes and other hard tissue. They are also an outstanding choice for industrial purposes and frozen sections.
Delivery information: Supplied in an ejector dispenser pack with a storage compartment to safely discard spent blades.