VWR® Magnetised pipette rack
供应商: VWR
VWR®
VWRU82007-220EA 0 CNY
VWRU82007-220
VWR® Magnetised pipette rack
吸管架
This durable blue ABS plastic storage rack features circular magnets for secure mounting to metal fume hoods, refrigerators, file cabinets, and more.
  • Four angled compartments accommodate 0,1 to 25 ml pipettes
  • Includes two movable attachments to hold pipetting devices