VWR®, Precision Digital Waterproof HACCP Thermometers with PT1000 Probe
供应商: VWR
VWR®
VWRI620-2644EA 0 CNY
VWRI620-2644 VWRI620-2646 VWRI620-2645
VWR®, Precision Digital Waterproof HACCP Thermometers with PT1000 Probe
温度计 数字温度计
Accurate measurements for laboratories, quality management, and monitoring of production processes. Application: Food (HACCP), medical/pharmaceutical science, chemistry, aquaristics, fish farming, aquaculture, and more.
  • Easy handling
  • Battery life time >6000 hours
  • Device and probe robust and waterproof
  • With PT1000, two-wire, isolated steam- and waterproof, permanently connected to device
  • Response time T90: Approx. 10
  • Can be recalibrated at 0 °C and scale
Accessories information: Compliance with HACCP and DIN EN 13485.
Delivery information: Device incl. probe, battery, calibration protocol and manual.