VWR®, Hard Rubber Spatulas
供应商: Avantor
VWR®
VWRMNPH/R-6EA 0 CNY
VWRMNPH/R-6 VWRMNPH/R-4 VWRMNPH/R-8
VWR®, Hard Rubber Spatulas
样品勺
These black rubber spatulas are ideal for mixing, weighing, cleaning, blending, and many others.
  • Hand-finished and hand-polished blades
  • Rust-proof, washable and sterilisable