VWR®EcoSense® pH10A, ph/°c meter, pen style
供应商: VWR
VWR®
VWRU74730-030EA 0 CNY
VWRU74730-030
VWR®EcoSense® pH10A, ph/°c meter, pen style
电化学仪器 多参数测量仪
The pH10A meter is the perfect instrument for economical spot sampling of pH and temperature in applications such as wastewater, surface water, aquaculture, hydroponics, pools, and education.
  • Automatic calibration and buffer recognition
  • 50-set memory (pH, temp, date and time stamp)
  • Replaceable single- or double-junction electrode
  • GLP functionality
  • 1-, 2-, or 3-point calibration
  • Hold feature locks stable readings on the display
  • >200 hour battery life with low battery indicator
This ultra-compact instrument even includes a graphic display with on-screen instructions. Available with single- or dual-junction electrodes (comes with single-junction electrode). No need to throw this pen-style pH instrument away when the sensor goes bad like some other products. Simply replace the electrode within minutes and keep testing.
Waterproof according to IP 67.
Warranty: 1 year
Delivery information: Supplied with 606110 pH electrode, batteries, and instruction manual.