These digital incubator provides an economical and space saving solution for microbiology or haematology applications.
- Digital controls
- Personal sized
The temperature can be freely adjusted in increments of 0,1 °C up to a maximum of 70 °C. Heating elements are incorporated into the housing base and walls to ensure temperature stability and uniformity. The door of the incubator has a transparent plexiglas® window to provide full visibility to samples. The housing and door frame are constructed entirely of epoxy coated mild steel for strength and durability.