Glass beads
供应商: Walter Stern
WALU100FEA 0 CNY
WALU100F
Glass beads
珠子 玻璃珠
These soda lime glass beads have a lower softening point (505 °C) than borosilicate glass.
  • Resistant to water, acid and chemicals
Diameter and spherical variation maximum is 10% of glass bead diameter. The temperature of transformation is Tg 440 °C, and the softening temperature is Tt 505 °C.
Packaging: One package contains approximately 2700 pieces and the pieces are delivered in a plastic jar.