Ready-to-use, 70% denatured ethanol (EtOH) solution is made to clean surfaces and equipments found in cleanroom areas within pharmaceutical, biotechnology and medical-device facilities and in the places of a non-pyrogenic alcohol solution is required.
- Gamma-irradiated to ensure sterility
- Evaporates completely, leaving no residue
Solution are made with Water-for-Injection (WFI) and filtered to 0,2 µm. Each lot undergoes a bacterial endotoxin test to ensure that the product meets or exceeds water-for-injection quality in endotoxin limits and each lot also undergoes a USP 14-day sterility test.
Certifications: EPA registered.
Delivery information: Each case is shipped with lot specific documentation which details QC, irradiation, sterility and bacterial endotoxin limits.
Packaging: Packaged in a individually double-bagged in a Class 100 cleanroom.