O-Rings for flat flanges
供应商: Chemglass
CGLACG-305-210EA 0 CNY
CGLACG-305-210 CGLACG-305-116
O-Rings for flat flanges
密封件 Sealing Rings
Viton O-ring is stocked in brown 75 durometer. Ideal for acids, aliphatic hydrocarbons, oxidising agents and all petroleum products.
  • Temperature range: –30 to 200° C
  • Low compression set and high temperature resistance
  • Especially good for vacuum service and low gas permeability