Adapters, expansion, cone on socket
供应商: Chemglass
CGLACG-1002-07EA 0 CNY
CGLACG-1002-07
Adapters, expansion, cone on socket
适配器 带圆锥形接地接头的适配器
Used to connect dissimilar standard taper joint sizes. Expansion adapter from small cone to large socket.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes