Adapters, reduction
供应商: Chemglass
CGLACG-1000-29EA 0 CNY
CGLACG-1000-44 CGLACG-1000-29
Adapters, reduction
适配器 带圆锥形接地接头的适配器
Used to connect dissimilar standard taper joint sizes. Reduction adapter from large cone at the base to small socket on the top.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes