Anti-MHC1 Mouse Monoclonal Antibody

供应商: Novus Biologicals
NOVUNB500-304EA 0 CNY
NOVUNB500-304
Anti-MHC1 Mouse Monoclonal Antibody
抗体
Mouse Monoclonal MHC Class I Antibody (MEM-E/02). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Primate.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: MHC1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR