Boc-D-propargylglycine 95%, 98% ee

供应商: Thermo Scientific
Warning

ACRO362850010EA 0 CNY
ACRO362850050 ACRO362850010
Boc-D-propargylglycine 95%, 98% ee
Boc-D-propargylglycine
分子式: C₁₀H₁₅NO₄
分子量: 213,233 g/mol
溶点: 93…98 °C
存储条件: Ambient
MDL编号: MFCD01320886
CAS号: 63039-46-3

现在订购

产品技术规格

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR