三氟乙酸 ≥99%, OR, Macron Fine Chemicals™
|
Danger
|
Synonyms:
TFA
MACR1942-57EA
0
CNY
MACR1942-57
MACR1942-59
三氟乙酸 ≥99%, OR, Macron Fine Chemicals™
三氟乙酸
分子式:
C₂HF₃O₂ 分子量: 114,02 g/mol 沸点: 71,8 °C (1013 hPa) 溶点: –15 °C 密度: 1,490 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00004169 CAS号: 76-05-1 EINECS: 200-929-3 UN号: 2699 ADR: 8,I Merck Index: 12,09812 |
产品技术规格
For Laboratory,Research,or Manufacturing Use | |
Assay (F₃CCOOH) | ≥99 % |
Appearance (colorless to light straw fuming liquid) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...