Stopcocks, straight bore, PTFE, SP Wilmad-LabGlass
供应商: SP Industries
SP Wilmad-LabGlass
LABGLG-9606T-102EA 0 CNY
LABGLG-9606T-102
Stopcocks, straight bore, PTFE, SP Wilmad-LabGlass
旋塞阀
Borosilicate glass.
Straight 1:5 taper PTFE stopcock with a threaded needle valve that extends through the bore of the plug, it can be used as a valve within the stopcock to control flow while using the stopcock for on-off control.