Fiber glass roving, PYREX®
供应商: Corning
PYREX®
CORN3950EA 0 CNY
CORN3950
Fiber glass roving, PYREX®
玻璃棉
This fiber glass is made from fine glass fibers with diameters controlled to approximately 0,008 mm. This makes the glass pliable and easy to use. It contains minimal heavy metals and is suitable for use as a filtering medium.
Manufactured from No. 9989 glass.
订购须知 This product is only sold in a case qty of 2,72 kg.
Packaging: Supplied in the form of roving approximately 2" in diameter by 22' long and packaged in 0,23 kg bags.