Parafilm for packaging, semi-transparent, thermoplastic
供应商: BEMIS COMPANY INC
BEMCPM996EA 0 CNY
BEMCPM996 BEMCPM999 BEMCPM998 BEMCPM992
Parafilm for packaging, semi-transparent, thermoplastic
密封件 密封膜
This film adheres quickly and firmly to itself, covering culture tube or flask mouths and sealing Petri dish edges.
  • Waterproof sheet interwound with paper to prevent adhering
  • Flexible, odourless and tasteless
  • Resistant to air, gases, alcohol, acids and alkalies
Stretches 200% at 21 °C and returns partially to its original size. At 82 to 93 °C , it softens enough to make an adhesive bond between papers, cloths, leathers, etc.
Delivery information: Supplied in dispenser cartons.