Craft blades, Slice®
供应商: Slice
Slice®
KNIFSLICECRAFTBLADEA 0 CNY
KNIFSLICECRAFTBLAD
Craft blades, Slice®
剃刀和刀片
These Slice® craft blades provide controlled, detailed cuts and a much safer finger-friendly® cutting edge.
  • Lasts up to 10× longer than steel blades
  • Straight edge with safer rounded tip
  • Does not conduct electricity
  • Never rusts, chemically inert, non-magnetic
Slice® craft blades are designed to work exclusively with Slice® handles for optimal safety and function.