Blades for cutters, Slice®
供应商: The Safety Knife Company
Slice®
KNIFSLICEREPLBLADEEA 0 CNY
KNIFSLICEREPLBLADE
Blades for cutters, Slice®
剃刀和刀片
These 100 percent zirconium oxide Slice® blades are compatible with Slice® Box Cutters, Pen Cutters, and the Mini Cutter.
  • Dual-sided
  • Lasts up to 10x longer than steel blades
  • Does not conduct electricity
  • Never rusts, chemically inert, non-magnetic
  • Safer blades compared to traditional steel
  • Hand sharpened
Slice ceramic blades require no oil coating or special maintenance.