VWR®, Tape dispenser
供应商: VWR
VWR®
VWRU89098-148EA 0 CNY
VWRU89098-148
VWR®, Tape dispenser
标签和胶带分配器
Dispenser is single-roll and designed with an adjustable core size to hold either a 12,7 m (41,7") long roll of tape on a 25,4 mm (1") core or a 54,9 m (180") long roll of tape on a 76,2 mm (3") core.
  • Heavy weighted base that provides increased stability and rests securely on any flat surface