VWR®, Tape and label dispenser
供应商: VWR
VWR®
VWRU89098-146EA 0 CNY
VWRU89098-146
VWR®, Tape and label dispenser
标签和胶带分配器
Dispenser for tapes and labels are handheld and can accommodates a maximum width of 29 mm.
  • Made of durable plastic
  • With two spacers and a plastic locking clip
  • Adjustable for rolls with 12,7 m (41,7") core and 12,7 mm (¹/₂"), 19 mm (³/₄") and 25 mm (1") widths