VWR® Talon® , Rod end connector
供应商: VWR
VWR® Talon®
VWRU60079-010EA 162.81 CNY
VWRU60079-010
VWR® Talon® , Rod end connector
搅拌棒
This nickel-plated zinc connector holds rods firmly at 90°.
  • Use this rod end connector when semi-permanent installations are required
  • Comes with two set screws and is precision bored for close fit with 13 mm (¹/₂") rods