VWR®, Staining Jars according to Hellendahl
供应商: VWR
VWR®
VWRU25460-907EA 0 CNY
VWRU25460-907
VWR®, Staining Jars according to Hellendahl
染色罐
PMP, with cover.
Holds 76×26 mm slides vertically. Eight grooves accommodate 16 slides back to back. Xylene can cause etching after 2 days at room temperature.