Acids and corrosives safety cabinets, lined, ChemCor®
供应商: Justrite
ChemCor®
JUSU8912022EA 0 CNY
JUSU8945822 JUSU8930822 JUSU8923222 JUSU8912022 JUSU8912222 JUSU8913222 JUSU8930222
Acids and corrosives safety cabinets, lined, ChemCor®
家具 储藏柜
An all epoxy baked-on powder-coat finish, inside and out, provides increased chemical resistance. Interior ChemCor® liner ensures complete corrosion resistance and protection. Unique, flame-coated thermoplastic liner provides superior protection against corrosion from drips and off-gasing from harsh chemicals.
  • 100% seamless liner protects exposed cabinet surfaces from rust and corrosion
  • Seam-free surface eliminates the need for rivets or fastners which risk corrosion from seep-through chemicals
  • Non-porous surface resists staining and cleans up easily
  • Three point stainless steel bullet self-latching system
  • U•Loc™ padlockable handle and Haz-Alert™ reflective warning labels
Cabinets are FM approved. Self-close models comply with OSHO, NFPA 30, NFPA 1 and International Fire Code™ regulations.
Colour: Blue