Tris-glycine buffer (10X), AccuGENE™
供应商: Lonza
AccuGENE®
LONZ50882EA 0 CNY
LONZ50882
Tris-glycine buffer (10X), AccuGENE™
缓冲(液) 电泳缓冲液 Transfer Buffers
AccuGENE™ electrophoresis buffers are prepared with high quality reagents and use 18 megOhm water. Products are filtered using a 0,45 µm filter.
  • Ready to use
  • Reliable, ensures consistency over lot-to-lot
  • Flexible