Accessories for VITEK® 2
供应商: bioMérieux
BMER21348EA 0 CNY
BMERV1200 BMER21348
Accessories for VITEK® 2
微生物学测试系统 抗生素试验
VITEK® 2 is an automated instrument for ID/AST testing. VITEK®2 uses unique ID and AST cards the size and shape of a playing card. Ready-to-use and low-cost, VITEK®2 ID cards offer a comprehensive menu of available tests. Based on this breakthrough innovation.
  • Ready to use
  • VITEK®2 can provide identification and susceptibility results in as little as 5 hours