Stopper, HDPE, KIMBLE®
供应商: DWK Life Sciences
KIMBLE®
KIML28160R-19EA 0 CNY
KIML28160R-19 SCERSP28160R-13 KIML28160R-9 KIML28160R-16
Stopper, HDPE, KIMBLE®
塞子 聚乙烯/低密度聚乙烯/高密度聚乙烯塞
HDPE, linear, ideal for flasks that are grounded to [ST] stopper dimensions.
  • Yellow and blue stopper with a closed bottom
  • Enlarged flange is designed to protect the neck if the glassware is tipped over