Procedure stations, PuriCare®
供应商: Labconco
PuriCare®
LBCP311621040EA 0 CNY
LBCP311621040
Procedure stations, PuriCare®
通风柜 层流柜
These PuriCare procedure station provides personnel, animal and environmental protection. It is specialised Class II biological safety cabinet that meets the product and personnel protection requirements of NSF 49. This stations is designed for conducting small animal research activities including cage changing, weighing, anesthesia delivery and surgical procedures.
  • Fully-closing, clear ¼" tempered safety glass sash with two sash handles; counterbalanced, anti-racking mechanism; and 10° slope
  • Electronic security lock
  • Filter monitoring system and electronically commutated motor (ECM) that automatically adjusts as filters load
  • Fluorescent lighting
  • Built-in interval or elapsed timer for experiment monitoring, fluorescent light or UV light control
  • Touchpad control on right-hand side post for manual activation of blower, light, timer, audible alarm mute and menu selection
  • With trace odour carbon postfilter
Construction: Work surface made of stainless steel 304, housing made from epoxy coated steel.