Anti-IgG F(c) Goat Polyclonal Antibody

供应商: Rockland Immunochemicals
ROCK612-1103EA 0 CNY
ROCK612-1103
Anti-IgG F(c) Goat Polyclonal Antibody
抗体
Secondary Goat Anti-IgG F(c) Reacts with Rat

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rat
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR