Anti-Ig Donkey Polyclonal Antibody (Biotin)

供应商: Cytiva
GEHERPN1004-2MLEA 0 CNY
GEHERPN1004-2ML
Anti-Ig Donkey Polyclonal Antibody (Biotin)
抗体
The antibodies are supplied with dilution guidelines for relevant applications.

  • Soluble in cold water and hot water

The antibodies are supplied with dilution guidelines for relevant applications and are stored at 2°C to 8°C.

Type: Secondary
Antigen: Rabbit Ig
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR